Order Entry
Italy
ContactUsLinkComponent
D-Gluconic acid 50% (w/w) in water
D-Gluconic acid 50% (w/w) in water
Certificates
undefined
D-Gluconic acid 50% (w/w) in water

Specification Test Results

  • Appearance:
    Clear yellow to brown-yellow Solution
  • Titration excess NaOH:
    49 to 55 %
  • Reducing sugars:
    ≤3 %
  • Titration Argentometric:
    ≥95.0 %

Specification Test Results

Related Information

Frequently Bought Together