Order Entry
Italy
ContactUsLinkComponent
Boron trifluoride dimethyl ether complex
Irritant
Health
Corrosive
Flammable
Boron trifluoride dimethyl ether complex
Certificates
undefined
Irritant
Health
Corrosive
Flammable
Boron trifluoride dimethyl ether complex

Specification Test Results

  • Appearance:
    Clear colorless to yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3070 to 1.3090 (20°C, 589 nm)
  • Assay:
    ≥59 % (BF3)

Specification Test Results

Related Information

Frequently Bought Together