



Petroleum spirit, 30…50 °C, EMSURE® for analysis, Supelco®
Catalog # 1.01786.2500
Supplier: Merck
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Solubility:(25 °C) insoluble
- Boiling Point:30 - 50 °C (1013 hPa)
- Explosion Limit:1.2 - 7.5 %(V)
- Density:0.65 g/cm3 (20 °C)
- Ignition Point:285 °C
Specification Test Results
About this item
:
Boiling Pt:30…50 °C (1013 hPa)
Density:0.65 g/cm³ (20 °C)
EINECS:265-151-9
ADR:3,I
:M-Clarity™ quality level = MQ300
Flash Pt:–40 °C
UN:1268
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849