Order Entry
Italy
ContactUsLinkComponent
Molecular sieve A4 (0.4 nm, 4 Å), rods ~ 1.6 mm (1/16"), Supelco®
Molecular sieve A4 (0.4 nm, 4 Å), rods ~ 1.6 mm (1/16"), Supelco®
Catalog # 1.05743.1000
Supplier:  Merck
CAS Number:  
Molecular sieve A4 (0.4 nm, 4 Å), rods ~ 1.6 mm (1/16"), Supelco®
Catalog # 1.05743.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Density:1.1…2.2 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:215-283-8
CAS Number:70955-01-0
 :1318-02-1
MDL Number:MFCD03457537