
Molecular sieve A4 (0.4 nm, 4 Å), powder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:70955-01-0
:1318-02-1
MDL Number:MFCD03457537
Density:1.1…2.2 g/cm³ (20 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together