



Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD02179311
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:66…69 °C(1.013 hPa)
Melting Pt:–95 °C
Density:0.660…0.668 g/cm³ (20 °C)
Flash Pt:–26 °C
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS07
:GHS08
:GHS02
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Autoignition Temperature:453 °F
- IR spectrum:complying
- Assay (GC):Min. 95.0 %
- Density (D 20/20):0.658 - 0.662
- Refractive index (n 20/D):1.373 - 1.377
- Non-volatile matter:Max. 0.0015 %
- Water (Karl Fischer):Max. 0.015 %
- Aromatic substances (as C₆H₆):Max. 0.005 %
- Vapor Pressure:~132 mmHg ( 20 °C)
- Explosion Limit:7.7 %
Specification Test Results
Frequently Bought Together