Palladium ≥99.9% (metals basis), wire, hard, Ø 0.05 mm
Catalog # 40730.G1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 m
- Seq:8362
- Pk Info:1 m×0.05 mm (l x Ø)
Specifications
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Merck Index:12,07121
Boiling Pt:3167 °C (1013 hPa)
Melting Pt:1555 °C
Density:12.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-115-6