Palladium ≥99.9% (metals basis), foil, Thickness: 1.0 mm
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
Merck Index:12,07121
Boiling Pt:3167 °C (1013 hPa)
Melting Pt:1555 °C
Density:12.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-115-6
Frequently Bought Together