Nickel ≥99.98% (metals basis), wire, hard, Ø 0.01 mm (0.0004 in)
Catalog # 40673.G1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 m
- Seq:8362
Specifications
About this item
CAS Number:7440-02-0
:
Formula:Ni
MDL Number:MFCD00011137
:58.69 g/mol
Merck Index:12,06582
Boiling Pt:3007 °C (1013 hPa)
Melting Pt:1453 °C
Density:8.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-111-4
UN:3089
ADR:4.1,III
:GHS08
:GHS07