Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iridium
Iridium ≥99.9% (metals basis), wire, Ø 0.075 mm (0.003 in)
Iridium ≥99.9% (metals basis), wire, Ø 0.075 mm (0.003 in)
Iridium ≥99.9% (metals basis), wire, Ø 0.075 mm (0.003 in)
44714.BS
:
Thermo Fisher Scientific
:
Iridium ≥99.9% (metals basis), wire, Ø 0.075 mm (0.003 in)
44714.BS
:
Thermo Fisher Scientific
:
Pk:
10 cm
Seq:
7600
:
:
192.22 g/mol
:
5300 °C (1013 hPa)
:
2450 °C
:
Ambient
:
231-095-9
:
7439-88-5
:
Ir
:
MFCD00011062