
Iron, EMSURE® 10 µm for analysis, reduced, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7439-89-6
:
Formula:Fe
MDL Number:MFCD00010999
:55.85 g/mol
Boiling Pt:3000 °C/1013 hPa
Melting Pt:1535 °C
Density:7.87 g/cm3 (20 °C)
EINECS:231-096-4
ADR:4.1,III
:M-Clarity™ quality level = MQ300
UN:3089
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Assay (cerimetric):≥ 99.5 %
- Substances insoluble in sulfuric acid:≤ 0.1 %
- Chloride (Cl):≤ 0.002 %
- Sulphide (S):≤ 0.002 %
- As (Arsenic):≤ 0.0002 %
- Pb (Lead):≤ 0.002 %
- Cu (Copper):≤ 0.002 %
- Mn (Manganese):≤ 0.002 %
- Zn (Zinc):≤ 0.002 %
Specification Test Results
Related Information
Frequently Bought Together