
Iron 99%, powder -70 mesh (<212 µm)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-89-6
:
Formula:Fe
MDL Number:MFCD00010999
:55.85 g/mol
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:1535 °C
Density:7,86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-096-4
ADR:4.2,II
:GHS02
Specification Test Results
- Appearance:Grey Powder
- Assay:≥98.5 %
- Copper (Cu):≤100 ppm
- Manganese (Mn):≤1000 ppm
- Nickel (Ni):≤500 ppm
- Sulfur (S):≤200 ppm
- Zinc (Zn):≤50 ppm
Specification Test Results
Frequently Bought Together