Order Entry
Italy
ContactUsLinkComponent
Iron 99+% (metals basis), wire, Ø 2.0 mm (0.08 in)
Iron 99+% (metals basis), wire, Ø 2.0 mm (0.08 in)
Catalog # 14246.H2
CAS Number:  
Iron 99+% (metals basis), wire, Ø 2.0 mm (0.08 in)
Catalog # 14246.H2
CAS Number:  

Specifications

  • Pk:
    50 m
  • Seq:
    8418

Specifications

About this item

 :
 :55.85 g/mol
Merck Index:13,05109
Boiling Pt:3000 °C (1013 hPa)
Melting Pt:1535 °C
Density:7.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-096-4
CAS Number:7439-89-6
Formula:Fe
MDL Number:MFCD00010999