
Cobalt ≥99.95% (metals basis), rods, Ø 5 mm (0.2 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-48-4
:
Formula:Co
MDL Number:MFCD00010935
:58.93 g/mol
Merck Index:12,02488
Boiling Pt:2870 °C (1013 hPa)
Melting Pt:1495 °C
Density:8.9 g/cm³ (20 °C)
EINECS:231-158-0
:GHS08
Storage Temperature:Ambient
Frequently Bought Together