Order Entry
Italy
ContactUsLinkComponent
 
Health
Cobalt ≥99.9% (metals basis), lumps typ. 1.5 cm cubes low oxygen, vacuum remelted
Catalog # 12956.22
CAS Number:  
Health
Cobalt ≥99.9% (metals basis), lumps typ. 1.5 cm cubes low oxygen, vacuum remelted
Catalog # 12956.22
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :58.93 g/mol
Merck Index:12,02488
Boiling Pt:2870 °C (1013 hPa)
Melting Pt:1495 °C
Density:8.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-158-0
 :GHS08
CAS Number:7440-48-4
Formula:Co
MDL Number:MFCD00010935