Order Entry
Italy
ContactUsLinkComponent
Cobalt ≥99.95% (metals basis), rods, Ø 12.5 mm (0.492 in)
Health
Cobalt ≥99.95% (metals basis), rods, Ø 12.5 mm (0.492 in)
Catalog # 00081.DH
CAS Number:  
Health
Cobalt ≥99.95% (metals basis), rods, Ø 12.5 mm (0.492 in)
Catalog # 00081.DH
CAS Number:  

Specifications

  • Pk:
    25 mm
  • Seq:
    1640

Specifications

About this item

 :
 :58.93 g/mol
Merck Index:12,02488
Boiling Pt:2870 °C (1013 hPa)
Melting Pt:1495 °C
Density:8.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-158-0
 :GHS08
CAS Number:7440-48-4
Formula:Co
MDL Number:MFCD00010935