Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vanadium
Vanadium ≥99.8% (metals basis), wire, Ø 0.075 mm (0.003 in)
Vanadium ≥99.8% (metals basis), wire, Ø 0.075 mm (0.003 in)
:
Thermo Fisher Scientific
Vanadium ≥99.8% (metals basis), wire, Ø 0.075 mm (0.003 in)
:
Thermo Fisher Scientific
:
7440-62-2
:
:
V
:
MFCD00011453
:
50.94 g/mol
:
12,10053
:
3477 °C (1013 hPa)
:
1896 °C
:
6.015 g/cm³ (20 °C)
:
Ambient
:
231-171-1
externalurl.pdf
msds_ALFA044108
Frequently Bought Together
Acetone, EMPARTA® ACS analytical reagent, Supelco®
: Merck
Dichloromethane stabilised, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Cyclohexane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.001 % H₂O)
: Merck
p-Xylene ≥99%, GPR RECTAPUR®
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
: VWR Chemicals
Toluene ≥99.7%, PESTINORM® for pesticide residue analysis
: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals