Niobium hard ≥99.8% (metals basis), rods, hard, Ø 19 mm (0.75 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-03-1
:
Formula:Nb
MDL Number:MFCD00011126
:92.91 g/mol
Boiling Pt:3700 °C (1013 hPa)
Melting Pt:2468 °C
Density:8.57 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-113-5
Frequently Bought Together