Order Entry
Italy
ContactUsLinkComponent
Zirconium ≥99.5% (metals basis excluding Hf), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Zirconium ≥99.5% (metals basis excluding Hf), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41659.KS
CAS Number:  
Zirconium ≥99.5% (metals basis excluding Hf), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 41659.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-67-7
 :
Formula:Zr
MDL Number:MFCD00011303
 :91.22 g/mol
Storage Temperature:Ambient
EINECS:231-176-9