Zirconium ≥99.8% (metals basis excluding Hf), lumps 3-6 mm (0.1-0.2 in) (max. 4% Hf)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-67-7
:
Formula:Zr
MDL Number:MFCD00011303
:91.22 g/mol
Storage Temperature:Ambient
EINECS:231-176-9
Related Information
Frequently Bought Together