Order Entry
Italy
ContactUsLinkComponent
Zirconium ≥99.2% (metals basis excluding Hf), plate (max. 4.5% Hf), Thickness: 3.175 mm (0.125 in)
Zirconium ≥99.2% (metals basis excluding Hf), plate (max. 4.5% Hf), Thickness: 3.175 mm (0.125 in)
Catalog # 10593.GH
CAS Number:  
Zirconium ≥99.2% (metals basis excluding Hf), plate (max. 4.5% Hf), Thickness: 3.175 mm (0.125 in)
Catalog # 10593.GH
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :91.22 g/mol
Storage Temperature:Ambient
EINECS:231-176-9
CAS Number:7440-67-7
Formula:Zr
MDL Number:MFCD00011303