Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 2.0 mm (0.08 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-67-7
:
Formula:Zr
MDL Number:MFCD00011303
:91.22 g/mol
Storage Temperature:Ambient
EINECS:231-176-9
Frequently Bought Together