Zirconium ≥99.95% (metals basis excluding Hf), wire (max. 0.25% Hf), Ø 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-67-7
:
Formula:Zr
MDL Number:MFCD00011303
:91.22 g/mol
Storage Temperature:Ambient
EINECS:231-176-9
Frequently Bought Together