
Titanium ≥99.5% (metals basis), powder -60+100 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-32-6
:
Formula:Ti
MDL Number:MFCD00011264
:47.88 g/mol
Boiling Pt:3277 °C (1013 hPa)
Melting Pt:1677 °C
Density:4.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-142-3
UN:2546
ADR:4.2,III
:GHS02
Related Information
Frequently Bought Together