Order Entry
Italy
ContactUsLinkComponent
Titanium ≥99.995% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Flammable
Titanium ≥99.995% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 43042.KS
CAS Number:  
Flammable
Titanium ≥99.995% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 43042.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :47.88 g/mol
Merck Index:12,09610
Boiling Pt:3262 °C (1013 hPa)
Melting Pt:1660 °C
Density:4.51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-142-3
 :GHS02
CAS Number:7440-32-6
Formula:Ti
MDL Number:MFCD00011264