Order Entry
Italy
ContactUsLinkComponent
Tellurium ≥99.9% (metals basis), ingot/button, Ø ∼38 mm (1.5 in), Thickness: 6 mm (0.24 in)
Toxic
Tellurium ≥99.9% (metals basis), ingot/button, Ø ∼38 mm (1.5 in), Thickness: 6 mm (0.24 in)
Catalog # 45108.KF
CAS Number:  
Toxic
Tellurium ≥99.9% (metals basis), ingot/button, Ø ∼38 mm (1.5 in), Thickness: 6 mm (0.24 in)
Catalog # 45108.KF
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:13494-80-9
 :
Formula:Te
MDL Number:MFCD00134062
 :127.6 g/mol
Merck Index:12,09273
Boiling Pt:990 °C (1013 hPa)
Melting Pt:450 °C
Density:8.83 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-813-4
UN:3284
ADR:6.1,III
 :GHS06