Bismuth ≥99.999% (metals basis), foil, Thickness: 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-69-9
:
Formula:Bi
MDL Number:MFCD00134033
:208.98 g/mol
Merck Index:12,01297
Boiling Pt:1490 °C (1013 hPa)
Melting Pt:271…272 °C
Density:9.78 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-177-4
Frequently Bought Together