0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Arsenic
Arsenic ≥99% (metals basis), powder -20 mesh
Arsenic ≥99% (metals basis), powder -20 mesh
10101.18
:
Thermo Fisher Scientific
:
Arsenic ≥99% (metals basis), powder -20 mesh
10101.18
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
50 g
Seq:
3368
:
:
74.92 g/mol
:
5.7 g/cm³ (20 °C)
:
Ambient
:
213-148-6
:
1558
:
6.1,II
:
GHS06
:
GHS08
:
7440-38-2
:
As
:
MFCD00085309