Inserisci ordine
Italy
ContactUsLinkComponent
 
Stagno ≥99.9% (metals basis), ingot/button, Ø ∼36 mm (1.4 in), Thickness: 8 mm (0.3 in)
Catalog # 45110.KF
CAS Number:  
Stagno ≥99.9% (metals basis), ingot/button, Ø ∼36 mm (1.4 in), Thickness: 8 mm (0.3 in)
Catalog # 45110.KF
CAS Number:  

Specifications

  • Conf:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-31-5
 :
Formula:Sn
MDL Number:MFCD00133862
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7,31 g/cm³ (25 °C)
 :118.71 g/mol
Storage Temperature:Ambiente
EINECS:231-141-8