Order Entry
Italy
ContactUsLinkComponent
Tin ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Tin ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40900.KS
CAS Number:  
Tin ≥99.995% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
Catalog # 40900.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:7440-31-5
 :
Formula:Sn
MDL Number:MFCD00133862
Merck Index:12,09587
Boiling Pt:2270 °C (1013 hPa)
Melting Pt:232 °C
Density:7.31 g/cm³ (25 °C)
 :118.71 g/mol
Storage Temperature:Ambient
EINECS:231-141-8