Order Entry
Italy
ContactUsLinkComponent
Carbon ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
  40830.KS
 :  
Carbon ≥99.999% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 3.18 mm (0.125 in)
  40830.KS
 :  
  • Pk:
    1
  • Seq:
    0001
 :
 :12.01 g/mol
 :13,01818
 :∼4000 °C (1013 hPa)
 :3727 °C
 :3.65 g/cm³ (20 °C)
 :Ambient
 :231-955-3
 :7440-44-0
 :C
 :MFCD00133992