Supelco® TraceCERT® Organic Reference Standard, Supelco 37 Component FAME Mix
Catalog # CRM47885
Supplier: Merck
CAS Number:
Specifications
- Concentration:In dichloromethane (varied conc.), ampule of 1 ml
- Pk:1 mL
- Standards:Certified reference material, TraceCERT®
- Description:Supelco 37 component FAME mix
- Storage conditions:–10...–25 °C
- Hazard class description:H315 - H319 - H336 - H351
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS08, GHS07
Caution: P202 - P261 - P264 - P302 + P352 - P305 + P351 + P338 - P308 + P313