Order Entry
Italy
ContactUsLinkComponent
Beakers, borosilicate glass, with graduation
Beakers, borosilicate glass, with graduation
Catalog # JONEBN.352
Supplier:  JONES FR
Supplier #: BN.352
undefined
Beakers, borosilicate glass, with graduation
Catalog # JONEBN.352
Supplier:  JONES FR
Supplier #: BN.352
Log in for availability
Select an Option
25 ml
Select an Option
With 10 graduations
Select an Option
02
Select an Option
1
About this item
These beakers are made of borosilicate glass in accordance with BS.1922. Each measure is government stamped.
  • Lighter, aids handling especially in the larger sizes
  • With permanent enamel graduations
  • Very good chemical and temperature resistance
The flat bottom allows for easier stirring on hotplates and homogenising. The calibration line spacing and accuracy are the same as traditional conical and bell measures.
Frequently Bought Together