Human recombinant PTX3 (from CHO cells)
ALEXALX201124C010
:
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Seq:01
- Protein/peptide name:PTX3
- Concentration:0,1 mg/ml
- Formulation:Liquid. In PBS containing 2% BSA and 0,1% sodium azide.
Produced in CHO cells. Human full-length PTX3.