Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Recombinant N-Acetyl-eglin C

Recombinant N-Acetyl-eglin C
Supplier:  
Certificates

0 Options Available Below

Product Details & Documents

Protease inhibitor

Eglin C was originally isolated from the leech Hirudo medicinalis . Recombinant N-acetyl-eglin C is a 70 amino acid peptide with identical biological activity to native eglin C. An effective inhibitor of chymotrypsin and subtilisin as well as of leukocyte elastase and cathepsin G.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ALEXALX201006MC01
  • ALEXALX201006MC05
  • ALEXALX201006M001
  • Protein/peptide name:
  • N-Acetyl-eglin C
  • N-Acetyl-eglin C
  • N-Acetyl-eglin C
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Purity:
  • >97% (HPLC)

Specifications

Product Family Options
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.