Human MMP2 proenzyme (from fibroblasts)
Catalog # ALEXALX200419C005
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:5 µG
- Source:Fibroblasts
- Species:Human
- Seq:01
- Protein/peptide name:MMP2 proenzyme
- Purity:>90% (SDS-PAGE, Western blot)
- Formulation:Liquid. In 50 mM TRIS-HCl, pH 7,0, containing 200 mM NaCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0,05% BRIJ 35 and 0,05% sodium azide.
- Purification:No other MMP contaminants are detectable
Specifications
About this item
Isolated from human rheumatoid synovial fibroblasts. Requires activation.