Anti-ITGB2 Mouse Monoclonal Antibody
Catalog # ALEXALX805043C100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Integrin Beta 2
- Antigen symbol:ITGB2
- Clonality:Monoclonal
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:01
- Form:Liquid. In PBS, pH 7,4, containing 15 mM sodium azide
- Antigen synonyms:LFA-1|CD18|MAC-1|integrin subunit beta 2|LAD|LCAMB|MFI7|MF17
- Concentration:1 mg/ml
- Purification:≥95% (SDS-PAGE)
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For Immunoprecipitation, Western Blot
Recognizes free human CD18 (beta2 integrin) not associated with CD11. Does not cross-react with pig CD18.
Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human