Anti-APRIL Rat Monoclonal Antibody
Catalog # ALEXALX804804C100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:A Proliferation-inducing Ligand
- Antigen symbol:APRIL
- Clonality:Monoclonal
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Seq:01
- Form:Liquid. In PBS containing 0,02% sodium azide
- Antigen synonyms:APRIL|TNFSF13|TALL2|TRDL-1|CD256|UNQ383/PRO715
- Concentration:1 mg/ml
- Purification:≥95% (SDS-PAGE)
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Host: Rat, Isotype: IgG2a
- For ELISA, Flow Cytometry, Immunocytochemistry
Recognizes human and mouse APRIL.
Type: Primary
Antigen: APRIL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human