Anti-CCL2 Mouse Monoclonal Antibody
Catalog # ALEXALX804465C100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen symbol:CCL2
- Clonality:Monoclonal
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:01
- Form:Liquid. In PBS containing 0,1% sodium azide
- Concentration:1 mg/ml
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Host: Mouse, Isotype: IgG1
- For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human CCL2.
Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human