Order Entry
Italy
ContactUsLinkComponent
 
Anti-AKT Mouse Monoclonal Antibody
Catalog # ALEXALX804401C100
Supplier:  Enzo Life Sciences
Anti-AKT Mouse Monoclonal Antibody
Catalog # ALEXALX804401C100
Supplier:  Enzo Life Sciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AKT
  • Antigen symbol:
    AKT
  • Clonality:
    Monoclonal
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    01
  • Form:
    Lyophilised from 1 ml of 2X PBS containing 0,09% sodium azide, PEG, and sucrose
  • Purification:
    Thiophilic adsorption and size exclusion chromatography purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat and dog Ser472/473/474-phosphorylated Akt (see technical note). Does not cross-react with dephosphorylated Akt or other unrelated serine-phosphorylated proteins.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: AKT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human