Anti-TLR9 Mouse Monoclonal Antibody
ALEXALX804364C100
- Antibody type:Primary
- Antigen name:Toll-like Receptor 9
- Antigen symbol:TLR9
- Clonality:Monoclonal
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Seq:01
- Form:Liquid. In PBS containing 0,05% BSA and 0,05% sodium azide
- Antigen synonyms:PDL1|PD-L1|CD289|B7H1|TLR9|Toll-like receptor 9|B7-H|Tlr9|PDCD1L1|PDCD1LG1
- Concentration:0,5 mg/ml
- Purification:Protein G-affinity purified
- Size:100 µg
- Pk:100 µG
Host: Mouse, Isotype: IgG1
- For Immunohistochemistry, Western Blot
Recognizes human, mouse, rat, horse, dog and monkey TLR9. Detects a band of ~120kDa by Western blot, may also detect a smaller band of TLR9 isoform B.
Type: Primary
Antigen: TLR9
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human