Anti-BIMS/EL/L Rat Monoclonal Antibody
Catalog # ALEXALX804351C100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen symbol:BIMS/EL/L
- Clonality:Monoclonal
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Seq:01
- Form:Liquid. In PBS containing 10 mM sodium azide
- Concentration:1 mg/ml
- Purification:Protein G-affinity purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Host: Rat, Isotype: IgG2a
- For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse and monkey BimL, BimEL and BimS.
Type: Primary
Antigen: BIMS/EL/L
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human