Anti-APAF 1 Rat Monoclonal Antibody
Catalog # ALEXALX804350C100
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:Apoptotic Peptidase Activating Factor 1
- Antigen symbol:APAF1
- Clonality:Monoclonal
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Seq:01
- Form:Liquid. Protein G purified antibody in PBS containing 0,02% sodium azide
- Antigen synonyms:DKFZp781B1145|CED4|KIAA0413
- Concentration:1 mg/ml
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Host: Rat, Isotype: IgG2a
- For ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Recognizes the CARD domain of human and mouse Apaf-1. Does not cross-react with rat Apaf-1.
Type: Primary
Antigen: APAF 1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human