Anti-FASLG Mouse Monoclonal Antibody
ALEXALX804273C100
- Antibody type:Primary
- Antigen name:Fas Ligand
- Antigen symbol:FASLG
- Clonality:Monoclonal
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Seq:01
- Neutralisation:Yes
- Form:Liquid. In PBS, pH 7,2, containing 1% BSA. Contains no preservatives
- Antigen synonyms:APT1LG1|APTL|TNFSF6|CD178|FASL|ALPS1B|CD95-L|CD95L
- Concentration:1 mg/ml
- Purification:≥95% (SDS-PAGE)
- Size:100 µg
- Pk:100 µG
Host: Mouse, Isotype: IgG1
Recognizes mouse FasL.
Type: Primary
Antigen: FASL
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human