Aliquat® 336 TG
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Methyltrialkylammonium chloride, Tricaprylmethylammonium chloride, Trioctylmethylammonium chloride, Methyltrioctylammonium chloride
CAS Number:63393-96-4
:
Formula:CH₃(CH₂)₇]₃NCH₃Cl
MDL Number:MFCD00011862
Density:0.89 g/cm³
Molecular Weight:404.17 g/mol
Flash Pt:77 °C
UN:2922
ADR:8 III
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to amber Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4655 to 1.4715 (20°C, 589 nm)
- Assay:112.0 to 117.2 (Quaternary value)
- Water:≤5 %
- Titration with HClO4:88.2 to 93.0 % (≤2.0 % (Tert.amine))
Specification Test Results
Frequently Bought Together