Order Entry
Italy
ContactUsLinkComponent
Organic reference standard, Glyphosate-FMOC, Dr. Ehrenstorfer
Organic reference standard, Glyphosate-FMOC, Dr. Ehrenstorfer
Catalog # EHERCA14051000
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Glyphosate-FMOC, Dr. Ehrenstorfer
Catalog # EHERCA14051000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standard
  • CAS No.:
    1373205-41-4
  • Description:
    Glyphosate-FMOC
  • Formula:
    C₁₈H₁₈NO₇P
  • Description:
    Glyphosate-FMOC

Specifications

About this item