Leishman's eosin-methylene blue stain for hematology
Catalog # A11644.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
EINECS:235-732-1
:GHS07
Storage Temperature:Ambient
CAS Number:12627-53-1
MDL Number:MFCD00131498