Order Entry
Italy
ContactUsLinkComponent
Malachite green phosphate assay kit
Malachite green phosphate assay kit
Catalog # ABNOKA1331
Supplier:  Abnova
CAS Number:  
undefined
Malachite green phosphate assay kit
Catalog # ABNOKA1331
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Malachite Green Phosphate Assay
  • Storage temperature:
    4 °C
  • Tests per kit:
    96 assays

Specifications

About this item

Malachite Green Phosphate Assay Kit provides a fast, reproducible, and non-radioactive method for measuring inorganic free phosphate in aqueous solution.

Delivery: The kit includes MG phosphate standard 1 M (100 µl), MG blue solution (1,8 ml×2), MG acidic solution (600 µl×2), 96-well solid plate (low volume) (2 plates) and 96-well cover sheet (2 sheets).
Caution: For research use only.