Photometric test kits for the determination of BOD₅ in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Coloured pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kits are evaluated in 16 mm (OD) tubes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Model:MANA985822
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):24
- Weight:232 g (including packaging)
IFU NANOCOLOR® Photometric Tube Test Kits for BOD5-TT.pdf IFU NANOCOLOR® Photometric Tube Tests, BOD₅.pdf IFU_Nanocolor Nutrient Mixture Plus with Nath IFU_Nanocolor Nutrient Mixture Without Nath