Anti-Profilin 1 Polyclonal Antibody
BSBTPB9313
- Antibody type:Primary
- Antigen symbol:Profilin 1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Polyclonal antibody for Profilin 1/PFN1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. Profilin 1/PFN1 information: Molecular Weight: 15054 MW; Subcellular Localization: Cytoplasm, cytoskeleton; Tissue Specificity: Expressed in epididymis (at protein level).
Type: Primary
Antigen: Profilin 1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat